Assay Details
| Gene Symbol: LOC4347312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756313.2 | XP_015611799.1 | 2 - 3 | 548 | 85 | |
| GenBank mRNA | AK062785.1 | - | 2 - 3 | 362 | 85 | |
More Information
Set Membership: |
|

LOC4347312
Genomic Map