Assay Details
| Gene Symbol: LOC4347313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755212.2 | XP_015610698.1 | 3 - 4 | 758 | 95 | |
| GenBank mRNA | AB080248.1 | - | 3 - 4 | 764 | 95 | |
| AK063940.1 | - | 3 - 4 | 529 | 95 | ||
| AK102698.1 | - | 1 - 2 | 137 | 95 | ||
More Information
Set Membership: |
|

LOC4347313
Genomic Map