Assay Details
| Gene Symbol: LOC4347319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756226.2 | XP_015611712.1 | 1 - 2 | 431 | 57 | |
| GenBank mRNA | AF402804.1 | - | 1 - 2 | 349 | 57 | |
| AK060223.1 | - | 1 - 2 | 346 | 57 | ||
| AK061838.1 | - | 1 - 2 | 348 | 57 | ||
| AK099489.1 | - | 1 - 2 | 349 | 57 | ||
More Information
Set Membership: |
|

LOC4347319
Genomic Map