Assay Details
| Gene Symbol: LOC4347381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755270.2 | XP_015610756.1 | 3 - 4 | 558 | 138 | |
| XM_015755271.2 | XP_015610757.1 | 3 - 4 | 561 | 138 | ||
| XM_015755273.2 | XP_015610759.1 | 2 - 3 | 459 | 138 | ||
| GenBank mRNA | AK121624.1 | - | 3 - 4 | 551 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015755270.2, XM_015755271.2, XM_015755273.2 transcripts.
Set Membership: |
|

LOC4347381
Genomic Map