Assay Details
| Gene Symbol: LOC4347381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755270.2 | XP_015610756.1 | 5 - 6 | 3710 | 79 | |
| XM_015755271.2 | XP_015610757.1 | 5 - 6 | 3713 | 79 | ||
| XM_015755273.2 | XP_015610759.1 | 4 - 5 | 3611 | 79 | ||
| GenBank mRNA | AK121624.1 | - | 5 - 6 | 3703 | 79 | |
More Information
Set Membership: |
|

LOC4347381
Genomic Map