Assay Details
| Gene Symbol: LOC4347508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756179.1 | XP_015611665.1 | 4 - 5 | 410 | 85 | |
| GenBank mRNA | AK061034.1 | - | 4 - 5 | 387 | 85 | |
| AK069451.1 | - | 4 - 5 | 367 | 85 | ||
More Information
Set Membership: |
|

LOC4347508
Genomic Map