Assay Details
| Gene Symbol: LOC4347889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756548.2 | XP_015612034.1 | 1 - 2 | 507 | 103 | |
| XM_015756549.2 | XP_015612035.1 | 1 - 2 | 507 | 103 | ||
| GenBank mRNA | AK065913.1 | - | 1 - 2 | 432 | 103 | |
More Information
Set Membership: |
|

LOC4347889
Genomic Map