Assay Details
| Gene Symbol: LOC4347889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756548.2 | XP_015612034.1 | 6 - 7 | 2972 | 83 | |
| XM_015756549.2 | XP_015612035.1 | 7 - 8 | 2867 | 83 | ||
| GenBank mRNA | AK065913.1 | - | 7 - 8 | 2793 | 83 | |
More Information
Set Membership: |
|

LOC4347889
Genomic Map