Assay Details
| Gene Symbol: LOC4348844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759364.2 | XP_015614850.1 | 8 - 9 | 951 | 83 | |
| GenBank mRNA | AK064778.1 | - | 8 - 9 | 912 | 83 | |
| AK104909.1 | - | 8 - 9 | 833 | 83 | ||
More Information
Set Membership: |
|

LOC4348844
Genomic Map