Assay Details
| Gene Symbol: LOC4348941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758684.2 | XP_015614170.1 | 1 - 2 | 745 | 69 | |
| XM_015758686.2 | XP_015614172.1 | 1 - 2 | 745 | 69 | ||
| GenBank mRNA | AK243136.1 | - | 1 - 2 | 716 | 69 | |
More Information
Set Membership: |
|

LOC4348941
Genomic Map