Assay Details
| Gene Symbol: LOC4348941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758684.2 | XP_015614170.1 | 9 - 10 | 2178 | 87 | |
| XM_015758686.2 | XP_015614172.1 | 9 - 10 | 2178 | 87 | ||
More Information
Set Membership: |
|

LOC4348941
Genomic Map