Assay Details
| Gene Symbol: LOC4348944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757500.2 | 18 - 19 | 3235 | 121 | ||
| XM_015757501.2 | 17 - 18 | 3082 | 121 | |||
| XM_015757502.2 | XP_015612988.1 | 17 - 18 | 3223 | 121 | ||
More Information
Set Membership: |
|

LOC4348944
Genomic Map