Assay Details
| Gene Symbol: LOC4348944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757500.2 | 24 - 25 | 4701 | 146 | ||
| XM_015757501.2 | 23 - 24 | 4548 | 146 | |||
| XM_015757502.2 | XP_015612988.1 | 23 - 24 | 4686 | 146 | ||
More Information
Set Membership: |
|

LOC4348944
Genomic Map