Assay Details
| Gene Symbol: LOC4349093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757280.2 | XP_015612766.1 | 10 - 11 | 1265 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4349093| Gene Symbol: LOC4349093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757280.2 | XP_015612766.1 | 10 - 11 | 1265 | 101 | |
Set Membership: |
|