Assay Details
| Gene Symbol: LOC4349093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757280.2 | XP_015612766.1 | 13 - 14 | 1525 | 117 | |
| GenBank mRNA | AK073197.1 | - | 12 - 13 | 1825 | 117 | |
More Information
Set Membership: |
|

LOC4349093
Genomic Map