Assay Details
| Gene Symbol: LOC4349762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761445.2 | XP_015616931.1 | 7 - 8 | 956 | 113 | |
| GenBank mRNA | AK065900.1 | - | 7 - 8 | 922 | 113 | |
| AK072412.1 | - | 7 - 8 | 922 | 113 | ||
More Information
Set Membership: |
|

LOC4349762
Genomic Map