Assay Details
| Gene Symbol: LOC4326136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765825.2 | XP_015621311.1 | 2 - 3 | 428 | 78 | |
| GenBank mRNA | AK103194.1 | - | 2 - 3 | 418 | 78 | |
More Information
Set Membership: |
|

LOC4326136
Genomic Map