Assay Details
| Gene Symbol: LOC4326136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765825.2 | XP_015621311.1 | 3 - 4 | 530 | 79 | |
| GenBank mRNA | AK101017.1 | - | 3 - 4 | 525 | 79 | |
| AK103194.1 | - | 3 - 4 | 520 | 79 | ||
More Information
Set Membership: |
|

LOC4326136
Genomic Map