Assay Details
| Gene Symbol: LOC4326136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765825.2 | XP_015621311.1 | 5 - 6 | 744 | 99 | |
| GenBank mRNA | AK101017.1 | - | 5 - 6 | 739 | 99 | |
| AK103194.1 | - | 5 - 6 | 734 | 99 | ||
More Information
Set Membership: |
|

LOC4326136
Genomic Map