Assay Details
| Gene Symbol: LOC4326074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787774.2 | XP_015643260.1 | 1 - 2 | 242 | 87 | |
| GenBank mRNA | AK059097.1 | - | 1 - 2 | 213 | 87 | |
| AK099387.1 | - | 1 - 2 | 212 | 87 | ||
More Information
Set Membership: |
|

LOC4326074
Genomic Map