Assay Details
| Gene Symbol: LOC4326074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787774.2 | XP_015643260.1 | 2 - 3 | 515 | 108 | |
| GenBank mRNA | AK059097.1 | - | 2 - 3 | 486 | 108 | |
| AK099387.1 | - | 2 - 3 | 485 | 108 | ||
More Information
Set Membership: |
|

LOC4326074
Genomic Map