Assay Details
| Gene Symbol: LOC4326074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787774.2 | XP_015643260.1 | 3 - 4 | 621 | 78 | |
| GenBank mRNA | AK059097.1 | - | 3 - 4 | 592 | 78 | |
| AK099387.1 | - | 3 - 4 | 591 | 78 | ||
More Information
Set Membership: |
|

LOC4326074
Genomic Map