Assay Details
| Gene Symbol: LOC4325211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766021.2 | XP_015621507.1 | 7 - 8 | 751 | 74 | |
| GenBank mRNA | AK060920.1 | - | 7 - 8 | 714 | 74 | |
| GQ848048.1 | - | 7 - 8 | 685 | 74 | ||
| JN944359.1 | - | 7 - 8 | 691 | 74 | ||
| KX302615.1 | - | 7 - 8 | 691 | 74 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC4325211
Genomic Map