Assay Details
| Gene Symbol: LOC4327303 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772933.2 | XP_015628419.1 | 1 - 2 | 524 | 68 | |
| XM_015772941.2 | XP_015628427.1 | 2 - 3 | 441 | 68 | ||
| GenBank mRNA | AK109125.1 | - | 1 - 2 | 271 | 68 | |
More Information
Set Membership: |
|

LOC4327303
Genomic Map