Assay Details
| Gene Symbol: LOC4327094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777379.2 | XP_015632865.1 | 11 - 12 | 2269 | 68 | |
| XM_015777387.2 | XP_015632873.1 | 11 - 12 | 2235 | 68 | ||
| XM_015777394.2 | XP_015632880.1 | 11 - 12 | 2226 | 68 | ||
| GenBank mRNA | AK100525.1 | - | 11 - 12 | 2271 | 68 | |
| AY332472.1 | - | 11 - 12 | 1854 | 68 | ||
More Information
Set Membership: |
|

LOC4327094
Genomic Map