Assay Details
| Gene Symbol: LOC4327094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777379.2 | XP_015632865.1 | 2 - 3 | 744 | 69 | |
| XM_015777387.2 | XP_015632873.1 | 2 - 3 | 728 | 69 | ||
| GenBank mRNA | AK100525.1 | - | 2 - 3 | 746 | 69 | |
| AY332472.1 | - | 2 - 3 | 347 | 69 | ||
More Information
Set Membership: |
|

LOC4327094
Genomic Map