Assay Details
| Gene Symbol: LOC4327094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777379.2 | XP_015632865.1 | 3 - 4 | 813 | 85 | |
| XM_015777387.2 | XP_015632873.1 | 3 - 4 | 797 | 85 | ||
| XM_015777394.2 | XP_015632880.1 | 3 - 4 | 770 | 85 | ||
| GenBank mRNA | AK100525.1 | - | 3 - 4 | 815 | 85 | |
| AY332472.1 | - | 3 - 4 | 416 | 85 | ||
More Information
Set Membership: |
|

LOC4327094
Genomic Map