Assay Details
| Gene Symbol: LOC4327094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777379.2 | XP_015632865.1 | 5 - 6 | 1063 | 71 | |
| XM_015777387.2 | XP_015632873.1 | 5 - 6 | 1047 | 71 | ||
| XM_015777394.2 | XP_015632880.1 | 5 - 6 | 1020 | 71 | ||
| GenBank mRNA | AK100525.1 | - | 5 - 6 | 1065 | 71 | |
| AY332472.1 | - | 5 - 6 | 666 | 71 | ||
More Information
Set Membership: |
|

LOC4327094
Genomic Map