Assay Details
| Gene Symbol: LOC4327094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777379.2 | XP_015632865.1 | 6 - 7 | 1146 | 88 | |
| XM_015777394.2 | XP_015632880.1 | 6 - 7 | 1103 | 88 | ||
More Information
Set Membership: |
|

LOC4327094
Genomic Map