Assay Details
| Gene Symbol: LOC4325263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777247.2 | XP_015632733.1 | 4 - 5 | 1237 | 67 | |
| GenBank mRNA | AK059088.1 | - | 4 - 5 | 1237 | 67 | |
| AK061118.1 | - | 4 - 5 | 977 | 67 | ||
| AK119598.1 | - | 4 - 5 | 974 | 67 | ||
| AK119642.1 | - | 4 - 5 | 977 | 67 | ||
More Information
Set Membership: |
|

LOC4325263
Genomic Map