Assay Details
| Gene Symbol: LOC4324283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762913.2 | XP_015618399.1 | 4 - 5 | 782 | 97 | |
| GenBank mRNA | AK318572.1 | - | 4 - 5 | 489 | 97 | |
More Information
Set Membership: |
|

LOC4324283
Genomic Map