Assay Details
| Gene Symbol: LOC4324283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762913.2 | XP_015618399.1 | 7 - 8 | 1145 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4324283| Gene Symbol: LOC4324283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762913.2 | XP_015618399.1 | 7 - 8 | 1145 | 75 | |
Set Membership: |
|