Assay Details
| Gene Symbol: LOC4326914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788431.2 | XP_015643917.1 | 1 - 2 | 265 | 122 | |
| GenBank mRNA | AK107209.1 | - | 1 - 2 | 265 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788431.2 transcripts.
Set Membership: |
|

LOC4326914
Genomic Map