Assay Details
| Gene Symbol: LOC4326914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788431.2 | XP_015643917.1 | 4 - 5 | 961 | 83 | |
| GenBank mRNA | AK107209.1 | - | 4 - 5 | 961 | 83 | |
| AY647458.1 | - | 4 - 5 | 910 | 83 | ||
More Information
Set Membership: |
|

LOC4326914
Genomic Map