Assay Details
| Gene Symbol: LOC4326914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788431.2 | XP_015643917.1 | 5 - 6 | 1134 | 67 | |
| GenBank mRNA | AY647458.1 | - | 5 - 6 | 1083 | 67 | |
More Information
Set Membership: |
|

LOC4326914
Genomic Map