Assay Details
| Gene Symbol: LOC4326917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765391.2 | XP_015620877.1 | 2 - 3 | 568 | 77 | |
| GenBank mRNA | AK065323.1 | - | 2 - 3 | 488 | 77 | |
| AK122127.1 | - | 2 - 3 | 488 | 77 | ||
| X78142.1 | - | 1 - 2 | 218 | 77 | ||
More Information
Set Membership: |
|

LOC4326917
Genomic Map