Assay Details
| Gene Symbol: LOC4325164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790457.2 | XP_015645943.1 | 3 - 4 | 1285 | 66 | |
| GenBank mRNA | AK106850.1 | - | 3 - 4 | 1285 | 66 | |
More Information
Set Membership: |
|

LOC4325164
Genomic Map