Assay Details
| Gene Symbol: LOC4326478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775959.2 | XP_015631445.1 | 14 - 15 | 1389 | 96 | |
| XM_015775967.2 | XP_015631453.1 | 14 - 15 | 1355 | 96 | ||
| XM_015775974.2 | XP_015631460.1 | 14 - 15 | 1352 | 96 | ||
| XM_015775983.2 | XP_015631469.1 | 14 - 15 | 1390 | 96 | ||
More Information
Set Membership: |
|

LOC4326478
Genomic Map