Assay Details
| Gene Symbol: LOC4326478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775959.2 | XP_015631445.1 | 17 - 18 | 1710 | 127 | |
| XM_015775967.2 | XP_015631453.1 | 17 - 18 | 1676 | 127 | ||
| XM_015775974.2 | XP_015631460.1 | 17 - 18 | 1673 | 127 | ||
More Information
Set Membership: |
|

LOC4326478
Genomic Map