Assay Details
| Gene Symbol: LOC4325444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765667.2 | XP_015621153.1 | 9 - 10 | 1673 | 62 | |
| GenBank mRNA | AK063767.1 | - | 6 - 7 | 957 | 62 | |
| AK073323.1 | - | 9 - 10 | 1559 | 62 | ||
| AK121501.1 | - | 9 - 10 | 1680 | 62 | ||
| BT129155.1 | - | 2 - 3 | 225 | 62 | ||
More Information
Set Membership: |
|

LOC4325444
Genomic Map