Assay Details
Gene Symbol: LOC4324038 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769655.2 | XP_015625141.1 | 2 - 3 | 445 | 103 | |
GenBank mRNA | AK102942.1 | - | 1 - 2 | 516 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
