Assay Details
| Gene Symbol: LOC4324038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769655.2 | XP_015625141.1 | 3 - 4 | 676 | 115 | |
| GenBank mRNA | AK102942.1 | - | 2 - 3 | 747 | 115 | |
More Information
Set Membership: |
|

LOC4324038
Genomic Map