Assay Details
| Gene Symbol: LOC4327806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758417.2 | XP_015613903.1 | 8 - 9 | 1413 | 113 | |
| XM_015758425.1 | XP_015613911.1 | 8 - 9 | 1317 | 113 | ||
| XM_015758430.2 | XP_015613916.1 | 8 - 9 | 1415 | 113 | ||
| XM_015758437.2 | XP_015613923.1 | 8 - 9 | 1415 | 113 | ||
| GenBank mRNA | AK060594.1 | - | 2 - 3 | 215 | 113 | |
More Information
Set Membership: |
|

LOC4327806
Genomic Map