Assay Details
| Gene Symbol: LOC4327806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758425.1 | XP_015613911.1 | 10 - 11 | 1601 | 87 | |
| XM_015758437.2 | XP_015613923.1 | 10 - 11 | 1699 | 87 | ||
| GenBank mRNA | AK060594.1 | - | 4 - 5 | 499 | 87 | |
More Information
Set Membership: |
|

LOC4327806
Genomic Map