Assay Details
| Gene Symbol: LOC4324449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757571.2 | XP_015613057.1 | 4 - 5 | 875 | 113 | |
| GenBank mRNA | AK103040.1 | - | 4 - 5 | 881 | 113 | |
More Information
Set Membership: |
|

LOC4324449
Genomic Map