Assay Details
| Gene Symbol: LOC4324449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757571.2 | XP_015613057.1 | 5 - 6 | 1213 | 63 | |
| GenBank mRNA | AK058846.1 | - | 1 - 2 | 283 | 63 | |
| AK103040.1 | - | 5 - 6 | 1219 | 63 | ||
| BT130633.1 | - | 1 - 2 | 233 | 63 | ||
More Information
Set Membership: |
|

LOC4324449
Genomic Map