Assay Details
| Gene Symbol: LOC4325917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766176.1 | XP_015621662.1 | 6 - 7 | 902 | 87 | |
| GenBank mRNA | AK069337.1 | - | 6 - 7 | 864 | 87 | |
| AK104932.1 | - | 6 - 7 | 620 | 87 | ||
More Information
Set Membership: |
|

LOC4325917
Genomic Map