Assay Details
| Gene Symbol: LOC4326245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766208.2 | XP_015621694.1 | 7 - 8 | 1692 | 95 | |
| GenBank mRNA | AK120308.1 | - | 7 - 8 | 1703 | 95 | |
More Information
Set Membership: |
|

LOC4326245
Genomic Map