Assay Details
| Gene Symbol: LOC4326245 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766208.2 | XP_015621694.1 | 9 - 10 | 2075 | 82 | |
| GenBank mRNA | AK120308.1 | - | 9 - 10 | 2086 | 82 | |
More Information
Set Membership: |
|

LOC4326245
Genomic Map