Assay Details
| Gene Symbol: LOC4327164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795005.2 | XP_015650491.1 | 1 - 2 | 790 | 124 | |
| GenBank mRNA | AK101948.1 | - | 1 - 2 | 791 | 124 | |
| AK106555.1 | - | 1 - 2 | 613 | 124 | ||
More Information
Set Membership: |
|

LOC4327164
Genomic Map